accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONCO

CAS RN 941717-02-8

IUPAC Name: 4-isocyanato-4-phenyloxane
Synonyms: 4-isocyanato-4-phenyloxane
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: HKKGHFQAYVPNAN-UHFFFAOYSA-N
SMILES: O=C=NC1(CCOCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results