CAS RN 941717-06-2
IUPAC Name:
4-[2-(bromomethyl)benzenesulfonyl]morpholine
Synonyms:
4-[2-(bromomethyl)benzenesulfonyl]morpholine
Molecular Weight (g/mol):
320.2
Molecular Formula:
C11H14BrNO3S
InChi Key:
VRRKTYLWVKMVJU-UHFFFAOYSA-N
SMILES:
BrCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
1
–
1
of
1
results
1
4-{[2-(Bromomethyl)phenyl]sulfonyl}morpholine, 90%, Thermo Scientific™
CAS: 941717-06-2 Molecular Formula: C11H14BrNO3S Molecular Weight (g/mol): 320.201 MDL Number: MFCD09879984 InChI Key: VRRKTYLWVKMVJU-UHFFFAOYSA-N Synonym: 4-2-bromomethyl phenyl sulfonyl morpholine,2-morpholin-4-yl sulphonyl benzyl bromide,4-2-bromomethyl benzenesulfonyl morpholine,4-2-bromomethyl phenyl sulphonyl morpholine,2-morpholin-4-yl sulfonyl benzyl bromide,4-2-bromomethyl benzene-1-sulfonyl morpholine PubChem CID: 24229771 IUPAC Name: 4-[2-(bromomethyl)phenyl]sulfonylmorpholine SMILES: C1COCCN1S(=O)(=O)C2=CC=CC=C2CBr