accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 943-89-5

IUPAC Name: (2E)-3-(4-methoxyphenyl)prop-2-enoic acid
Synonyms: p-methoxy-cinnamic acid cinnamic acid, P-methoxy- P-methoxycinnamic acid (trans) methoxycinnamic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: AFDXODALSZRGIH-QPJJXVBHSA-N
SMILES: COC1=CC=C(\C=C\C(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results