accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HONNNCl

CAS RN 94361-06-5

IUPAC Name: 2-(4-chlorophenyl)-3-cyclopropyl-1-(1H-1,2,4-triazol-1-yl)butan-2-ol
Synonyms: cyproconazole
Molecular Weight (g/mol): 291.78
Molecular Formula: C15H18ClN3O
InChi Key: UFNOUKDBUJZYDE-UHFFFAOYNA-N
SMILES: CC(C1CC1)C(O)(CN1C=NC=N1)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Physical Form 
  • (2)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results