accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FFOHOFF

CAS RN 944-43-4

IUPAC Name: 4-amino-2,3,5,6-tetrafluorobenzoic acid
Synonyms: 4-amino-2,3,5,6-tetrafluorobenzoic acid
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: WTNSXWSOTDBWOR-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(C(O)=O)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results