accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNNH3CNH2

CAS RN 944401-55-2

IUPAC Name: 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Synonyms: 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Molecular Weight (g/mol): 235.09
Molecular Formula: C11H18BN3O2
InChi Key: SYJMHOBGFXCKRG-UHFFFAOYSA-N
SMILES: CC1=NC(N)=NC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results