accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNFFFNH2

CAS RN 944401-57-4

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 288.08
Molecular Formula: C12H16BF3N2O2
InChi Key: AHNBKJSRXQDYEO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(N)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results