accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONNNOOFFFFF

CAS RN 944450-81-1

IUPAC Name: tert-butyl 4-{4-[(2,3,4,5,6-pentafluorophenoxy)carbonyl]pyridin-2-yl}piperazine-1-carboxylate
Synonyms: tert-butyl 4-[4-(2,3,4,5,6-pentafluorophenoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol): 473.4
Molecular Formula: C21H20F5N3O4
InChi Key: XHATYMDMSNXMKQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=CC(=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results