accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 944710-34-3

H3CCH3CH3PHNPCH3CH3H3CH3CCH3CH3CH3CH3H3C

CAS RN 944710-34-3

IUPAC Name: bis[2-(di-tert-butylphosphanyl)ethyl]amine
Synonyms: bis[2-(di-tert-butylphosphanyl)ethyl]amine
Molecular Weight (g/mol): 361.54
Molecular Formula: C20H45NP2
InChi Key: MJYKYLNPIGDVEF-UHFFFAOYSA-N
SMILES: CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)