accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 945717-09-9

CHNCl

CAS RN 945717-09-9

IUPAC Name: 2-chloro-4-ethynylpyridine
Synonyms: 2-chloro-4-ethynylpyridine
Molecular Weight (g/mol): 137.57
Molecular Formula: C7H4ClN
InChi Key: DLEWWBKXDRBGRF-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results