accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 946-65-6

IUPAC Name: 3-[(trifluoromethyl)sulfanyl]benzoate
Synonyms: 3-[(trifluoromethyl)sulfanyl]benzoate
Molecular Weight (g/mol): 221.17
Molecular Formula: C8H4F3O2S
InChi Key: IVGAPIVNQABETQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results