accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 946-80-5

IUPAC Name: (benzyloxy)benzene
Synonyms: benzyl phenyl ether
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: BOTNYLSAWDQNEX-UHFFFAOYSA-N
SMILES: C(OC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)