CAS RN 94602-04-7
IUPAC Name:
4-ethoxy-3-nitropyridine hydrochloride
Synonyms:
4-ethoxy-3-nitropyridine hydrochloride
Molecular Weight (g/mol):
204.61
Molecular Formula:
C7H9ClN2O3
InChi Key:
HNMWMLXQJBWURC-UHFFFAOYSA-N
SMILES:
Cl.CCOC1=C(C=NC=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Ethoxy-3-nitropyridine hydrochloride, 97%, Thermo Scientific™
CAS: 94602-04-7 Molecular Formula: C7H9ClN2O3 Molecular Weight (g/mol): 204.61 MDL Number: MFCD01075663 InChI Key: HNMWMLXQJBWURC-UHFFFAOYSA-N Synonym: 4-ethoxy-3-nitropyridine hydrochloride,3-nitro-4-ethoxypyridine hydrochloride,4-ethoxy-3-nitro-pyridine hydrochloride,4-ethoxy-3-nitropyridine hydrogen chloride,pyridine, 4-ethoxy-3-nitro-, monohydrochloride,4-ethoxy-3-nitropyridinehydrochloride,pubchem6674,labotest-bb lt00454779,4-ethoxy-3-nitropyridine hcl,ksc496g1h PubChem CID: 12407497 IUPAC Name: 4-ethoxy-3-nitropyridine;hydrochloride SMILES: CCOC1=C(C=NC=C1)[N+](=O)[O-].Cl