accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 946168-04-3

BOOH3CCH3H3CH3CCH

CAS RN 946168-04-3

IUPAC Name: 2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 228.1
Molecular Formula: C14H17BO2
InChi Key: SJDFLZSEQGKSDJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results