accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 946607-10-9

H3CCH3H3CNNH3CCH3H3CCl

CAS RN 946607-10-9

IUPAC Name: 1,3-di-tert-butyl-1H-1,3-benzodiazol-3-ium chloride
Synonyms: 1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride
Molecular Weight (g/mol): 266.81
Molecular Formula: C15H23ClN2
InChi Key: CMQDFXSMZLANNA-UHFFFAOYSA-M
SMILES: [Cl-].CC(C)(C)N1C=[N+](C2=CC=CC=C12)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results