accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONH2FFFF

CAS RN 946784-69-6

IUPAC Name: 4-(4-fluorophenoxy)-2-(trifluoromethyl)aniline
Synonyms: 4-(4-fluorophenoxy)-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 271.22
Molecular Formula: C13H9F4NO
InChi Key: JYXYSTKHJJWGRL-UHFFFAOYSA-N
SMILES: NC1=C(C=C(OC2=CC=C(F)C=C2)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results