accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 947-42-2

OHSiHO

CAS RN 947-42-2

IUPAC Name: diphenylsilanediol
Synonyms: diphenylsilanediol
Molecular Weight (g/mol): 216.31
Molecular Formula: C12H12O2Si
InChi Key: OLLFKUHHDPMQFR-UHFFFAOYSA-N
SMILES: O[Si](O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results