accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONN

CAS RN 947-92-2

IUPAC Name: N-cyclohexyl-N-nitrosocyclohexanamine
Synonyms: N-nitrosodicyclohexylamine
Molecular Weight (g/mol): 210.32
Molecular Formula: C12H22N2O
InChi Key: CYMNKXLBEMEABS-UHFFFAOYSA-N
SMILES: O=NN(C1CCCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results