accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OOHHNOO

CAS RN 94744-50-0

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylpropanoic acid
Molecular Weight (g/mol): 325.36
Molecular Formula: C19H19NO4
InChi Key: HOZZVEPRYYCBTO-UHFFFAOYSA-N
SMILES: CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results