accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 94790-37-1

IUPAC Name: (1H-1,2,3-benzotriazol-1-yl)[bis(dimethylamino)methylidene]oxidanium
Synonyms: HBTU
Molecular Weight (g/mol): 234.28
Molecular Formula: C11H16N5O
InChi Key: CLZISMQKJZCZDN-UHFFFAOYSA-N
SMILES: CN(C)C(=[O+]N1N=NC2=CC=CC=C12)N(C)C
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results