accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-03-4

IUPAC Name: 5-chloro-2-methoxyaniline
Synonyms: 5-chloro-2-methoxyaniline
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: WBSMIPLNPSCJFS-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results