accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3C

CAS RN 95-47-6

IUPAC Name: 1,2-xylene
Synonyms: ortho-xylene o-xylol ccris 905 xylenes (isomer mixture)
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (7)
  • (1)
  • (3)
Solvent or Matrix 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)