accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HO

CAS RN 95-48-7

IUPAC Name: 2-methylphenol
Synonyms: o-cresol o-toluol orthocresol (substance) potassium salt 2-cresol o-cresylic acid sodium salt 2-cresol ammonium salt 2-cresol 0.1% w/w benzene ccris 646 o-kresol
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (7)
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (7)
Physical Form 
  • (7)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (7)
  • (7)
  • (3)