accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-53-4

IUPAC Name: 2-methylaniline
Synonyms: O-toluidine (ar)-isomer 2-toluidine lithium salt 2-toluidine ccris 597 o-toluidyna rcra waste number U328 2-toluidine, (ar)- (ar)-2-toluidine 2-toluidine, (ar)-isomer
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: RNVCVTLRINQCPJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)