accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2H2N

CAS RN 95-54-5

IUPAC Name: benzene-1,2-diamine
Synonyms: o-phenylenediamine phenylene-1,2-dimaine CI oxidation base 16 PODA 2-phenylene diamine ccris 508 o-fenylendiamin C.I. oxidation base 16 1,2-fenylendiamin orthamine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: GEYOCULIXLDCMW-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (6)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)