accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClCH3

CAS RN 95-66-9

IUPAC Name: 1-chloro-2,4-dimethylbenzene
Synonyms: MXYLENE4chloro
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: UIEVCEQLNUHDIF-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results