accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-68-1

IUPAC Name: 2,4-dimethylaniline
Synonyms: 2,4-dimethylaniline m-xylidine meta-xylidine ccris 254 m-4-xylidine asym-m-xylidene
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: CZZZABOKJQXEBO-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(C)=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results