accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3Cl

CAS RN 95-72-7

IUPAC Name: 2-chloro-1,4-dimethylbenzene
Synonyms: 2-chloro-1,4-dimethylbenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: KZNRNQGTVRTDPN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results