accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-76-1

IUPAC Name: 3,4-dichloroaniline
Synonyms: 3,4-dichloroaniline C6H5cl2N m.p-dichloroaniline
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: SDYWXFYBZPNOFX-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (6)
  • (5)
Boiling Point 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (6)
  • (5)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)