accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-78-3

IUPAC Name: 2,5-dimethylaniline
Synonyms: 2,5-xylidine p-dimethylaniline 2,5-xylidene para-xylidine p-xylidine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: VOWZNBNDMFLQGM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(N)=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)