accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-79-4

IUPAC Name: 5-chloro-2-methylaniline
Synonyms: 5-chloro-O-toluidine
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: WRZOMWDJOLIVQP-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1N
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)