accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-80-7

CH3NH2H2N

CAS RN 95-80-7

IUPAC Name: 4-methylbenzene-1,3-diamine
Synonyms: developer T benzofur MT renal MD 2,4-tolamine developer mtd developer dbj nako tmt CI oxidation base 35 3H-labeled 2,4-diaminotoluene pelagol J
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: VOZKAJLKRJDJLL-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)