accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95-82-9

NH2ClCl

CAS RN 95-82-9

IUPAC Name: 2,5-dichloroaniline
Synonyms: 2,5-dichloroaniline scarlet base ciba I fast scarlet base 2JS scarlet base ngg fast scarlet mdc base natasol scarlet GG salt stabamine scarlet GG diazo fast scarlet GG azobase dca scarlet base irga I (free base)
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: AVYGCQXNNJPXSS-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)