accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClH2N

CAS RN 95-83-0

IUPAC Name: 4-chlorobenzene-1,2-diamine
Synonyms: 4-chloro-1,2-benzenediamine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: BXIXXXYDDJVHDL-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1N
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)