accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClHO

CAS RN 95-88-5

IUPAC Name: 4-chlorobenzene-1,3-diol
Synonyms: 4-chlororesorcinol
Molecular Weight (g/mol): 144.55
Molecular Formula: C6H5ClO2
InChi Key: JQVAPEJNIZULEK-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C(O)=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)