accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NON(R)O(R)OH(R)HOFF

CAS RN 95058-81-4

IUPAC Name: 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: gemcitabine gamcitabine gemtro GEO gemzar gemcin DFdCyd gemcitabinum gemcitabina gemcitabina [inn-spanish]
Molecular Weight (g/mol): 263.2
Molecular Formula: C9H11F2N3O4
InChi Key: SDUQYLNIPVEERB-QPPQHZFASA-N
SMILES: NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)