accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 950603-21-1

IUPAC Name: 1-(thian-4-yl)methanamine hydrochloride
Synonyms: 1-(thian-4-yl)methanamine hydrochloride
Molecular Weight (g/mol): 167.7
Molecular Formula: C6H14ClNS
InChi Key: DREBXEFIPXBLAP-UHFFFAOYSA-N
SMILES: Cl.NCC1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results