accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 950603-35-7

IUPAC Name: 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Synonyms: 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Molecular Weight (g/mol): 281.25
Molecular Formula: C9H10F3N3O2S
InChi Key: PWQMNLMMMCREOV-UHFFFAOYSA-N
SMILES: OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results