accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95067-27-9

IUPAC Name: methyl 2-acetamido-3-nitrobenzoate
Synonyms: methyl 2-acetamido-3-nitrobenzoate
Molecular Weight (g/mol): 238.2
Molecular Formula: C10H10N2O5
InChi Key: PHFRWNCHBPVTJN-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(NC(C)=O)C(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results