accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(S)(R)O(R)NHOHNOOHO

CAS RN 951-78-0

IUPAC Name: 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: deoxyuridine
Molecular Weight (g/mol): 228.2
Molecular Formula: C9H12N2O5
InChi Key: MXHRCPNRJAMMIM-UHFFFAOYNA-N
SMILES: OCC1OC(CC1O)N1C=CC(=O)NC1=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)