accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOOH3COH3C

CAS RN 951-82-6

IUPAC Name: 2-(3,4,5-trimethoxyphenyl)acetic acid
Synonyms: (3,4,5-trimethoxyphenyl)acetic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C11H14O5
InChi Key: DDSJXCGGOXKGSJ-UHFFFAOYSA-N
SMILES: COC1=CC(CC(O)=O)=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results