accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOOHO

CAS RN 95124-68-8

IUPAC Name: 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoic acid
Synonyms: 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoic acid
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H8N2O3
InChi Key: DTKAUYQBZSOHRJ-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)