accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95167-41-2

CH3NCH3NHNOHO(Z)

CAS RN 95167-41-2

IUPAC Name: N'-[(Z)-[4-(diethylamino)phenyl]methylidene]-4-hydroxybenzohydrazide
Synonyms: N'-[(Z)-[4-(diethylamino)phenyl]methylidene]-4-hydroxybenzohydrazide
Molecular Weight (g/mol): 311.39
Molecular Formula: C18H21N3O2
InChi Key: WLKOCYWYAWBGKY-UYRXBGFRSA-N
SMILES: CCN(CC)C1=CC=C(\C=N/NC(=O)C2=CC=C(O)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results