accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 952182-03-5

IUPAC Name: N-methyloxetan-3-amine
Synonyms: N-methyloxetan-3-amine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: JUIXJPRSYHSLHK-UHFFFAOYSA-N
SMILES: CNC1COC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results