accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNCH3

CAS RN 953040-54-5

IUPAC Name: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole
Synonyms: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
Molecular Weight (g/mol): 207.08
Molecular Formula: C11H18BNO2
InChi Key: SYAMJGBBLFTQTK-UHFFFAOYSA-N
SMILES: CN1C=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results