accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 953408-82-7

OSOOCl

CAS RN 953408-82-7

IUPAC Name: 2,3-dihydro-1-benzofuran-7-sulfonyl chloride
Synonyms: 2,3-dihydro-1-benzofuran-7-sulfonyl chloride
Molecular Weight (g/mol): 218.65
Molecular Formula: C8H7ClO3S
InChi Key: UNECAYDZDLGSLP-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC2=C1OCC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)