accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 953408-88-3

IUPAC Name: 4-methyl-2-(pyridin-4-yl)-1,3-thiazole-5-carbonyl chloride hydrochloride
Synonyms: 4-methyl-2-(pyridin-4-yl)-1,3-thiazole-5-carbonyl chloride hydrochloride
Molecular Weight (g/mol): 275.15
Molecular Formula: C10H8Cl2N2OS
InChi Key: XERFFSPGBNKEDT-UHFFFAOYSA-N
SMILES: Cl.CC1=C(SC(=N1)C1=CC=NC=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results