accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 953780-78-4

IUPAC Name: 3-methylmorpholine hydrochloride
Synonyms: 3-methylmorpholine hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: MSOCQCWIEBVSLF-UHFFFAOYNA-N
SMILES: Cl.CC1COCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results