accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNHHON(E/Z)

CAS RN 95649-37-9

IUPAC Name: N-[amino(3H-indol-3-ylidene)methyl]hydroxylamine
Synonyms: N-[amino(indol-3-ylidene)methyl]hydroxylamine
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: DRCFXQBNDIRJCT-UHFFFAOYSA-N
SMILES: NC(NO)=C1C=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results