accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClNHOOHO(E/Z)

CAS RN 95695-47-9

IUPAC Name: 3-[(2,4-dichlorophenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(2,4-dichlorophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 260.07
Molecular Formula: C10H7Cl2NO3
InChi Key: WFWKZRKAECLVSE-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=C(Cl)C=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results